This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06632915_s1
See other HDAC3 GE Assays ›
Gene Symbol:
HDAC3
Gene Name:
histone deacetylase 3
Gene Aliases:
HD3, RPD3, RPD3-2
Chromosome Location:
Chr.5: 141620876 - 141636860 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06632915_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003883.3 NP_003874.2 7 603 95
XM_011537697.2 XP_011535999.1 6 455 95
GenBank mRNA AF005482.1 - 5 619 95
AF039703.1 - 7 559 95
AY429537.1 - 3 171 95
AY429538.1 - 5 286 95
BC000614.1 - 7 592 3345755 95
BM461585.1 - 7 593 95
U66914.1 - 7 548 95
U75696.1 - 6 586 95
U75697.1 - 7 592 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011537697.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HDAC3

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
positive regulation of protein phosphorylation
transcription, DNA-templated
protein deacetylation
circadian rhythm
negative regulation of myotube differentiation
chromatin modification
regulation of protein stability
positive regulation of TOR signaling
positive regulation of transcription factor import into nucleus
negative regulation of apoptotic process
cellular lipid metabolic process
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of JNK cascade
spindle assembly
histone H3 deacetylation
cellular response to fluid shear stress
chromatin binding
transcription corepressor activity
histone deacetylase activity
protein binding
transcription factor binding
enzyme binding
cyclin binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
NF-kappaB binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0