This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06633243_s1
See other CDHR5 GE Assays ›
Gene Symbol:
CDHR5
Gene Name:
cadherin related family member 5
Gene Aliases:
MLPCDH, MU-PCDH, MUCDHL, MUPCDH
Chromosome Location:
Chr.11: 616565 - 625067 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06633243_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDHR5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001171968.1 NP_001165439.1 1 111 91
NM_021924.4 NP_068743.2 1 111 91
NM_031264.3 NP_112554.2 1 111 91
GenBank mRNA AF258675.1 - 2 267 90
AF258676.1 - 2 267 90
AF258677.1 - 2 168 90
AF258678.1 - 1 210 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF258675.1, AF258676.1, NM_001171968.1, NM_021924.4, NM_031264.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDHR5

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
cell differentiation
regulation of microvillus length
intermicrovillar adhesion
calcium ion binding
protein binding
beta-catenin binding
cell adhesion molecule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rzthw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0