This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06635280_s1
See other CEL GE Assays ›
Gene Symbol:
CEL
Gene Name:
carboxyl ester lipase
Gene Aliases:
BAL, BSDL, BSSL, CELL, CEase, FAP, FAPP, LIPA, MODY8
Chromosome Location:
Chr.9: 133061978 - 133071863 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06635280_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001807.4 NP_001798.2 11 2316 86
GenBank mRNA AK299965.1 - 12 2128 86
BC042510.1 - 12 2199 5187959 86
BE045879.1 - 1 86
M85201.1 - 12 2271 86
S70517.1 - 1 127 86
X54457.1 - 12 2348 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CEL

Panther Classification:

Molecular Function -

cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
triglyceride metabolic process
cholesterol catabolic process
neuron cell-cell adhesion
synapse assembly
fatty acid catabolic process
protein esterification
pancreatic juice secretion
intestinal cholesterol absorption
lipid digestion
intestinal lipid catabolic process
modulation of synaptic transmission
catalytic activity
sterol esterase activity
triglyceride lipase activity
receptor activity
protein binding
heparin binding
hydrolase activity
neurexin family protein binding
acylglycerol lipase activity
carboxylic ester hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z47m5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0