This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06635463_s1
See other CRAT GE Assays ›
Gene Symbol:
CRAT
Gene Name:
carnitine O-acetyltransferase
Gene Aliases:
CAT1
Chromosome Location:
Chr.9: 129094794 - 129110791 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06635463_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000755.3 NP_000746.2 2 452 84
NM_001257363.1 NP_001244292.1 3 551 84
XM_005251708.3 XP_005251765.1 4 614 84
XM_017014274.1 XP_016869763.1 3 619 84
XM_017014275.1 XP_016869764.1 2 272 84
GenBank mRNA AK313931.1 - 3 463 84
AL523652.3 - 2 84
BC000723.2 - 2 391 3504259 84
BI460020.1 - 3 527 84
BI752361.1 - 2 446 84
BQ068230.1 - 476 84
BT006801.1 - 2 170 84
X78706.1 - 2 165 84
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DQ655951.1 - 1 Chr. 9 - 129107874 - 129109276 [+] on Build GRCh38 84
GenBankmRNA DQ655950.1 - 1 Chr. 9 - 129107874 - 129109276 [+] on Build GRCh38 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CRAT

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

transport
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA oxidase
carnitine O-acetyltransferase activity
receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5h84w:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0