This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06635469_s1
See other CRAT GE Assays ›
Gene Symbol:
CRAT
Gene Name:
carnitine O-acetyltransferase
Gene Aliases:
CAT1
Chromosome Location:
Chr.9: 129094794 - 129110791 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06635469_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257363.1 NP_001244292.1 2 336 59
XM_005251708.3 XP_005251765.1 2 328 59
XM_017014274.1 XP_016869763.1 2 328 59
GenBank mRNA AK313931.1 - 2 248 59
BI460020.1 - 2 312 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001257363.1, XM_005251708.3, XM_017014274.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

transport
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA oxidase
carnitine O-acetyltransferase activity
receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0