This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06636212_s1
See other MGST1 GE Assays ›
Gene Symbol:
MGST1
Gene Name:
microsomal glutathione S-transferase 1
Gene Aliases:
GST12, MGST, MGST-I
Chromosome Location:
Chr.12: 16347142 - 16377189 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06636212_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MGST1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260511.1 NP_001247440.1 2 159 106
NM_001260512.1 NP_001247441.1 2 166 106
NM_001267598.1 NP_001254527.1 2 166 106
NM_020300.4 NP_064696.1 2 166 106
NM_145764.2 NP_665707.1 2 149 106
NM_145791.2 NP_665734.1 2 219 106
NM_145792.2 NP_665735.1 2 141 106
NR_048545.1 2 166 106
NR_048547.1 2 219 106
XM_006719079.1 XP_006719142.1 2 157 106
XM_011520673.1 XP_011518975.1 2 157 106
XM_011520674.2 XP_011518976.1 2 157 106
GenBank mRNA AA314967.1 - 2 219 106
AK291148.1 - 2 155 106
AV655242.1 - 2 200 106
BC005923.1 - 2 134 4297300 106
BE568125.1 - 2 130 105
BG569388.1 - 2 150 106
BG617746.1 - 2 167 106
BG778603.1 - 2 167 106
BT006982.1 - 1 76 106
CR407629.1 - 1 76 106
H49331.1 - 2 164 106
J03746.1 - 2 149 106
R86750.1 - 2 141 106

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MGST1

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
xenobiotic metabolic process
response to organonitrogen compound
response to lipopolysaccharide
Leydig cell differentiation
response to drug
oxidation-reduction process
protein homotrimerization
cellular response to lipid hydroperoxide
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
glutathione peroxidase activity
protein binding
protein homodimerization activity
glutathione binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0