This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06636344_g1
See other PTPN11 GE Assays ›
Gene Symbol:
PTPN11
Gene Name:
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases:
BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2
Chromosome Location:
Chr.12: 112418732 - 112509913 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06636344_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002834.3 NP_002825.3 10 1542 114
NM_080601.1 NP_542168.1 10 1542 114
XM_006719526.2 XP_006719589.1 10 1320 114
XM_011538613.2 XP_011536915.1 10 1356 114
XM_017019722.1 XP_016875211.1 10 1359 114
GenBank mRNA AK289854.1 - 10 1320 114
BC008692.1 - 10 1365 3872007 114
BT007106.1 - 9 1162 114
D13540.1 - 10 1318 114
L03535.1 - 10 1275 114
L07527.1 - 10 1315 114
L08807.1 - 10 1360 114
S78088.1 - 1 58 114
X70766.1 - 10 1291 114

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PTPN11

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

DNA damage checkpoint
activation of MAPK activity
triglyceride metabolic process
epidermal growth factor receptor signaling pathway
integrin-mediated signaling pathway
axonogenesis
brain development
heart development
fibroblast growth factor receptor signaling pathway
hormone-mediated signaling pathway
regulation of phosphatidylinositol 3-kinase signaling
cerebellar cortex formation
platelet activation
platelet formation
T cell costimulation
microvillus organization
abortive mitotic cell cycle
regulation of cell adhesion mediated by integrin
negative regulation of cell adhesion mediated by integrin
multicellular organism growth
organ growth
peptidyl-tyrosine dephosphorylation
megakaryocyte development
phosphatidylinositol-3-phosphate biosynthetic process
atrioventricular canal development
ERBB signaling pathway
regulation of multicellular organism growth
hormone metabolic process
glucose homeostasis
regulation of protein complex assembly
positive regulation of mitotic cell cycle
negative regulation of insulin secretion
regulation of protein export from nucleus
phosphatidylinositol phosphorylation
positive regulation of hormone secretion
platelet-derived growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
ephrin receptor signaling pathway
phosphatidylinositol-mediated signaling
multicellular organismal reproductive process
genitalia development
inner ear development
homeostasis of number of cells within a tissue
leukocyte migration
negative regulation of cortisol secretion
Bergmann glial cell differentiation
negative regulation of growth hormone secretion
face morphogenesis
regulation of type I interferon-mediated signaling pathway
intestinal epithelial cell migration
positive regulation of ERK1 and ERK2 cascade
positive regulation of glucose import in response to insulin stimulus
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
non-membrane spanning protein tyrosine phosphatase activity
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
1-phosphatidylinositol-3-kinase activity
protein domain specific binding
receptor tyrosine kinase binding
D1 dopamine receptor binding
phospholipase binding
insulin receptor substrate binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
peptide hormone receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0