This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06636362_s1
See other P2RX4 GE Assays ›
Gene Symbol:
P2RX4
Gene Name:
purinergic receptor P2X 4
Gene Aliases:
P2X4, P2X4R
Chromosome Location:
Chr.12: 121209861 - 121234106 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06636362_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256796.1 NP_001243725.1 9 1169 86
NM_001261397.1 NP_001248326.1 8 1040 86
NM_001261398.1 NP_001248327.1 8 1121 86
NM_002560.2 NP_002551.2 8 1121 86
NR_046372.1 9 1159 86
NR_046373.1 7 973 86
XM_011538416.2 XP_011536718.1 4 484 86
GenBank mRNA AF000234.1 - 8 717 86
AF012903.1 - 7 603 86
AK225604.1 - 8 817 86
AK293599.1 - 5 611 86
AK293660.1 - 7 756 86
AK297704.1 - 7 705 86
AK301080.1 - 8 805 86
AK308549.1 - 9 891 86
AK308868.1 - 8 1812 86
BC017458.1 - 8 850 86
BC033826.1 - 8 841 5216449 86
BT019738.1 - 8 817 86
BT019739.1 - 8 736 86
DQ851491.1 - 4 335 86
U83993.1 - 8 1126 86
Y07684.1 - 8 844 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2RX4

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

tissue homeostasis
regulation of sodium ion transport
transport
signal transduction
blood coagulation
regulation of blood pressure
positive regulation of calcium ion transport into cytosol
negative regulation of cardiac muscle hypertrophy
neuronal action potential
sensory perception of pain
positive regulation of prostaglandin secretion
response to ATP
ion transmembrane transport
response to fluid shear stress
purinergic nucleotide receptor signaling pathway
endothelial cell activation
positive regulation of nitric oxide biosynthetic process
positive regulation of calcium-mediated signaling
protein homooligomerization
membrane depolarization
positive regulation of calcium ion transport
regulation of cardiac muscle contraction
relaxation of cardiac muscle
cellular response to ATP
apoptotic signaling pathway
cation transmembrane transport
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
receptor binding
copper ion binding
protein binding
ATP binding
drug binding
zinc ion binding
protein homodimerization activity
cadherin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0