This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06636442_s1
See other RARG GE Assays ›
Gene Symbol:
RARG
Gene Name:
retinoic acid receptor gamma
Gene Aliases:
NR1B3, RARC
Chromosome Location:
Chr.12: 53210566 - 53232256 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06636442_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RARG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042728.2 NP_001036193.1 1 349 55
NM_001243732.1 NP_001230661.1 1 349 55
GenBank mRNA AK297277.1 - 1 349 55
AK297801.1 - 1 195 55
AK301057.1 - 1 316 55
AK316259.1 - 1 73 55
CO394652.1 - 1 306 55
DC294855.1 - 1 285 55
HQ692829.1 - 1 36 55
M32074.1 - 1 97 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neural tube closure
glandular epithelial cell development
retinal pigment epithelium development
growth plate cartilage chondrocyte growth
transcription initiation from RNA polymerase II promoter
positive regulation of cell proliferation
negative regulation of cell proliferation
regulation of cell size
anterior/posterior pattern specification
embryonic camera-type eye development
regulation of myelination
negative regulation of chondrocyte differentiation
response to retinoic acid
embryonic hindlimb morphogenesis
multicellular organism growth
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of programmed cell death
steroid hormone mediated signaling pathway
regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
embryonic eye morphogenesis
retinoic acid receptor signaling pathway
canonical Wnt signaling pathway
face development
trachea cartilage development
prostate gland epithelium morphogenesis
Harderian gland development
cellular response to retinoic acid
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
retinoic acid receptor activity
protein binding
zinc ion binding
retinoid X receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0