This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06636730_s1
See other IL17D GE Assays ›
Gene Symbol:
IL17D
Gene Name:
interleukin 17D
Gene Aliases:
IL-17D
Chromosome Location:
Chr.13: 20702775 - 20723100 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06636730_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL17D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138284.1 NP_612141.1 2 289 58
XM_005266421.3 XP_005266478.1 2 733 58
GenBank mRNA AF458062.1 - 2 239 58
AF479775.1 - 1 181 58
AY078238.1 - 2 289 58
AY359113.1 - 1 181 58
BC036243.1 - 2 292 5301608 58

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IL17D

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
cell surface receptor signaling pathway
cytokine activity
cytokine receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline