This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06637123_g1
See other PPM1A GE Assays ›
Gene Symbol:
PPM1A
Gene Name:
protein phosphatase, Mg2+/Mn2+ dependent 1A
Gene Aliases:
PP2C-ALPHA, PP2CA, PP2Calpha
Chromosome Location:
Chr.14: 60245752 - 60299087 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06637123_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPM1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021003.4 NP_066283.1 2 601 70
NM_177951.2 NP_808820.1 3 703 70
NM_177952.2 NP_808821.2 2 466 70
XM_005267779.1 XP_005267836.1 3 548 70
XM_005267781.1 XP_005267838.1 3 706 70
XM_011536872.1 XP_011535174.1 5 968 70
XM_011536876.1 XP_011535178.1 4 825 70
XM_011536877.1 XP_011535179.1 5 868 70
XM_011536878.1 XP_011535180.1 4 791 70
XM_011536879.2 XP_011535181.1 4 660 70
XM_011536880.2 XP_011535182.1 5 714 70
XM_011536881.2 XP_011535183.1 3 574 70
XM_011536882.2 XP_011535184.1 2 243 70
XM_011536883.1 XP_011535185.1 3 657 70
XM_011536884.1 XP_011535186.1 2 214 70
XM_017021381.1 XP_016876870.1 5 1838 70
XM_017021382.1 XP_016876871.1 7 1088 70
XM_017021383.1 XP_016876872.1 4 724 70
XM_017021384.1 XP_016876873.1 4 416 70
XM_017021385.1 XP_016876874.1 3 691 70
XM_017021386.1 XP_016876875.1 3 454 70
XM_017021387.1 XP_016876876.1 4 700 70
GenBank mRNA AB451247.1 - 1 151 70
AB451371.1 - 1 151 70
AF070670.1 - 3 510 70
AK097843.1 - 2 70
AK308225.1 - 2 330 70
AK313337.1 - 3 548 70
AY236965.1 - 2 231 70
BC026691.1 - 2 454 3847638 70
BC063243.1 - 2 384 6011486 70
DA439296.1 - 3 503 70
S87759.1 - 2 508 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPM1A

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein dephosphorylation
N-terminal protein myristoylation
cell cycle arrest
negative regulation of SMAD protein complex assembly
Wnt signaling pathway
dephosphorylation
positive regulation of Wnt signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
peptidyl-threonine dephosphorylation
negative regulation of NF-kappaB import into nucleus
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of transcription, DNA-templated
positive regulation of protein export from nucleus
cellular response to transforming growth factor beta stimulus
magnesium ion binding
protein serine/threonine phosphatase activity
signal transducer activity
protein binding
manganese ion binding
calmodulin-dependent protein phosphatase activity
R-SMAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0