This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06637478_s1
See other CIB2 GE Assays ›
Gene Symbol:
CIB2
Gene Name:
calcium and integrin binding family member 2
Gene Aliases:
DFNB48, KIP2, USH1J
Chromosome Location:
Chr.15: 78104606 - 78131976 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06637478_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CIB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271888.1 NP_001258817.1 5 872 84
NM_001271889.1 NP_001258818.1 4 760 84
NM_001301224.1 NP_001288153.1 5 922 84
NM_006383.3 NP_006374.1 6 907 84
NR_125435.1 5 795 84
XM_006720374.2 XP_006720437.1 7 732 84
GenBank mRNA AB012955.1 - 6 593 84
BC033108.1 - 5 872 4841632 84
BC047381.1 - 6 835 5287072 84
BC057292.1 - 6 835 84
BE907622.1 - 152 84
BI523557.1 - 3 307 84
BM908035.1 - 534 84
BU556614.1 - 4 658 84
CN371149.1 - 2 171 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CIB2

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cytosolic calcium ion concentration
photoreceptor cell maintenance
calcium ion homeostasis
cellular response to ATP
magnesium ion binding
integrin binding
calcium ion binding
protein binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline