This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06640517_g1
See other PPP4R3B GE Assays ›
Gene Symbol:
PPP4R3B
Gene Name:
protein phosphatase 4 regulatory subunit 3B
Gene Aliases:
FLFL2, PP4R3B, PSY2, SMEK2, smk1
Chromosome Location:
Chr.2: 55547292 - 55617757 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06640517_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP4R3B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001122964.2 NP_001116436.2 17 5361 111
NM_001282850.1 NP_001269779.1 16 5265 111
NM_020463.3 NP_065196.1 15 5106 111
XM_005264442.2 XP_005264499.1 17 5340 111
XM_005264444.2 XP_005264501.1 16 5202 111
XM_005264445.2 XP_005264502.1 16 5181 111
XM_005264446.2 XP_005264503.1 15 5085 111
XM_005264447.2 XP_005264504.1 16 4737 111
XM_017004532.1 XP_016860021.1 16 4716 111
XM_017004533.1 XP_016860022.1 15 4641 111
XM_017004534.1 XP_016860023.1 15 4620 111
XM_017004535.1 XP_016860024.1 14 4482 111
XM_017004536.1 XP_016860025.1 14 4461 111
XM_017004537.1 XP_016860026.1 18 5552 111
XM_017004538.1 XP_016860027.1 18 5464 111
XM_017004539.1 XP_016860028.1 17 5348 111
XM_017004540.1 XP_016860029.1 17 5347 111
GenBank mRNA BE535214.1 - 1 106 111

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP4R3B

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s)

protein dephosphorylation
regulation of lipid metabolic process
positive regulation of gluconeogenesis

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0