This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06645290_s1
See other SUMF1 GE Assays ›
Gene Symbol:
SUMF1
Gene Name:
sulfatase modifying factor 1
Gene Aliases:
AAPA3037, FGE, UNQ3037
Chromosome Location:
Chr.3: 4034714 - 4467282 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06645290_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SUMF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011533624.2 XP_011531926.1 9 1134 93
XM_017006252.1 XP_016861741.1 8 1074 93
XM_017006253.1 XP_016861742.1 8 1056 93
XM_017006254.1 XP_016861743.1 9 1134 93
GenBank mRNA AY358092.1 - 9 1123 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
post-translational protein modification
oxidation-reduction process
oxidoreductase activity
protein homodimerization activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0