This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06645665_s1
See other NELFA GE Assays ›
Gene Symbol:
NELFA
Gene Name:
negative elongation factor complex member A
Gene Aliases:
NELF-A, P/OKcl.15, WHSC2
Chromosome Location:
Chr.4: 1982714 - 2009972 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06645665_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NELFA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005663.4 NP_005654.3 1 409 71
XM_017008589.1 XP_016864078.1 1 1146 71
GenBank mRNA AB044549.1 - 196 71
AF101434.1 - 1 138 71
AK001304.1 - 1 196 71
AK094040.1 - 1 141 71
AK126056.1 - 1 1284 71
AK222853.1 - 1 146 71
BC002764.2 - 1 149 3630594 71
DB082393.1 - 1 409 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK094040.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NELFA

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
multicellular organism development
negative regulation of transcription elongation from RNA polymerase II promoter
positive regulation of viral transcription
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline