This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06652535_g1
See other DPY30 GE Assays ›
Gene Symbol:
DPY30
Gene Name:
dpy-30, histone methyltransferase complex regulatory subunit
Gene Aliases:
Cps25, HDPY-30, Saf19
Chromosome Location:
Chr.2: 32011649 - 32039840 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06652535_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DPY30
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321209.1 NP_001308138.1 4 319 79
NM_001321210.1 NP_001308139.1 4 319 79
NM_032574.3 NP_115963.1 4 459 79
NR_135585.1 4 312 79
XM_006712117.3 XP_006712180.1 4 324 79
XM_006712118.3 XP_006712181.1 4 324 79
XM_011533133.2 XP_011531435.1 4 392 79
XM_017005111.1 XP_016860600.1 4 324 79
GenBank mRNA AF226998.1 - 4 378 79
AV725116.1 - 4 250 79
BC015970.1 - 4 281 4081996 79
BG940527.1 - 4 267 79
BI752326.1 - 4 280 79
BM505380.1 - 2 79
BQ276899.1 - 319 79

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DPY30

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing at telomere
transcription, DNA-templated
endosomal transport
histone H3-K4 methylation
protein binding
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline