This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06748892_g1
See other CNBP GE Assays ›
Gene Symbol:
CNBP
Gene Name:
CCHC-type zinc finger nucleic acid binding protein
Gene Aliases:
CNBP1, DM2, PROMM, RNF163, ZCCHC22, ZNF9
Chromosome Location:
Chr.3: 129167815 - 129183967 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06748892_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127193.1 NP_001120665.1 3 - 4 424 61
NM_001127195.1 NP_001120667.1 3 - 4 403 61
GenBank mRNA BT019613.1 - 2 - 3 218 61
DQ091187.1 - 3 - 4 356 61
DQ092366.1 - 3 - 4 335 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cholesterol biosynthetic process
positive regulation of cell proliferation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
single-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
single-stranded RNA binding
protein binding
zinc ion binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0