This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07286639_g1
See other P2RX2 GE Assays ›
Gene Symbol:
P2RX2
Gene Name:
purinergic receptor P2X 2
Gene Aliases:
DFNA41, P2X2
Chromosome Location:
Chr.12: 132618780 - 132623678 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07286639_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282164.1 NP_001269093.1 4 - 5 482 83
NM_001282165.1 NP_001269094.1 5 - 6 589 83
NM_012226.4 NP_036358.2 3 - 4 373 83
NM_016318.3 NP_057402.1 4 - 5 517 83
NM_170682.3 NP_733782.1 5 - 6 589 83
NM_170683.3 NP_733783.1 5 - 6 589 83
NM_174872.2 NP_777361.1 3 - 4 313 83
NM_174873.2 NP_777362.1 5 - 6 589 83
XM_005266154.3 XP_005266211.1 5 - 6 573 83
XM_005266155.4 XP_005266212.1 5 - 6 583 83
XM_005266156.4 XP_005266213.1 5 - 6 583 83
XM_011534786.2 XP_011533088.1 4 - 5 501 83
XM_017019035.1 XP_016874524.1 5 - 6 583 83
GenBank mRNA AF109387.1 - 5 - 6 516 83
AF109388.1 - 5 - 6 516 83
AF190822.1 - 5 - 6 552 83
AF190823.1 - 5 - 6 552 83
AF190824.1 - 4 - 5 480 83
AF190825.1 - 5 - 6 552 83
AF260426.1 - 5 - 6 589 83
AF260427.1 - 5 - 6 552 83
AF260428.1 - 3 - 4 276 83
AF260429.1 - 3 - 4 336 83
AY346374.1 - 4 - 5 445 83
BC109200.2 - 5 - 6 552 40005772 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2RX2

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
detection of hypoxic conditions in blood by carotid body chemoreceptor signaling
neuromuscular synaptic transmission
neuromuscular junction development
blood coagulation
sensory perception of sound
response to carbohydrate
positive regulation of calcium ion transport into cytosol
urinary bladder smooth muscle contraction
peristalsis
response to ATP
purinergic nucleotide receptor signaling pathway
behavioral response to pain
skeletal muscle fiber development
positive regulation of calcium-mediated signaling
sensory perception of taste
protein homooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ATP binding
ligand-gated ion channel activity
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0