This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07286640_g1
See other P2RX2 GE Assays ›
Gene Symbol:
P2RX2
Gene Name:
purinergic receptor P2X 2
Gene Aliases:
DFNA41, P2X2
Chromosome Location:
Chr.12: 132618780 - 132623678 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07286640_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282164.1 NP_001269093.1 5 - 6 571 68
NM_001282165.1 NP_001269094.1 6 - 7 678 68
NM_012226.4 NP_036358.2 4 - 5 462 68
NM_016318.3 NP_057402.1 5 - 6 606 68
NM_170682.3 NP_733782.1 6 - 7 678 68
NM_170683.3 NP_733783.1 6 - 7 678 68
NM_174872.2 NP_777361.1 4 - 5 402 68
NM_174873.2 NP_777362.1 6 - 7 678 68
XM_005266154.3 XP_005266211.1 6 - 7 662 68
XM_005266155.4 XP_005266212.1 6 - 7 672 68
XM_005266156.4 XP_005266213.1 6 - 7 672 68
XM_011534786.2 XP_011533088.1 5 - 6 590 68
XM_017019035.1 XP_016874524.1 6 - 7 672 68
GenBank mRNA AF109387.1 - 6 - 7 605 68
AF109388.1 - 6 - 7 605 68
AF190822.1 - 6 - 7 641 68
AF190823.1 - 6 - 7 641 68
AF190824.1 - 5 - 6 569 68
AF190825.1 - 6 - 7 641 68
AF260426.1 - 6 - 7 678 68
AF260427.1 - 6 - 7 641 68
AF260428.1 - 4 - 5 365 68
AF260429.1 - 4 - 5 425 68
AY346374.1 - 5 - 6 534 68
BC109200.2 - 6 - 7 641 40005772 68

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2RX2

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
detection of hypoxic conditions in blood by carotid body chemoreceptor signaling
neuromuscular synaptic transmission
neuromuscular junction development
blood coagulation
sensory perception of sound
response to carbohydrate
positive regulation of calcium ion transport into cytosol
urinary bladder smooth muscle contraction
peristalsis
response to ATP
purinergic nucleotide receptor signaling pathway
behavioral response to pain
skeletal muscle fiber development
positive regulation of calcium-mediated signaling
sensory perception of taste
protein homooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ATP binding
ligand-gated ion channel activity
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0