This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07286836_m1
See other NTMT1 GE Assays ›
Gene Symbol:
NTMT1
Gene Name:
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases:
AD-003, C9orf32, HOMT1A, METTL11A, NRMT, NTM1A
Chromosome Location:
Chr.9: 129608884 - 129636742 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07286836_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NTMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286796.1 NP_001273725.1 3 - 4 769 94
NM_001286797.1 NP_001273726.1 3 - 4 582 94
NM_001286798.1 NP_001273727.1 3 - 4 643 94
NM_001286799.1 NP_001273728.1 3 - 4 572 94
NM_001286802.1 NP_001273731.1 2 - 3 427 94
NM_001286803.1 NP_001273732.1 2 - 3 401 94
NM_014064.3 NP_054783.2 3 - 4 617 94
XM_005251939.3 XP_005251996.1 3 - 4 558 94
XM_017014642.1 XP_016870131.1 3 - 4 475 94
GenBank mRNA AF110776.1 - 3 - 4 560 94
AI802417.1 - 1 - 2 394 94
AK292332.1 - 3 - 4 619 94
AK313829.1 - 3 - 4 607 94
BC001396.2 - 3 - 4 596 3050940 94
BC033234.1 - 3 - 4 571 5426447 94
BI083998.1 - 3 - 4 582 94
BM009113.1 - 2 - 3 354 94
BM554138.1 - 3 - 4 572 94
HY058177.1 - 2 - 3 423 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NTMT1

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

spindle organization
chromosome segregation
N-terminal peptidyl-alanine methylation
N-terminal peptidyl-alanine trimethylation
N-terminal peptidyl-proline dimethylation
N-terminal peptidyl-serine dimethylation
N-terminal peptidyl-serine trimethylation
protein methyltransferase activity
N-terminal protein N-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline