This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07286930_m1
See other ZNF302 GE Assays ›
Gene Symbol:
ZNF302
Gene Name:
zinc finger protein 302
Gene Aliases:
HSD16, MST154, MSTP154, ZNF135L, ZNF140L, ZNF327
Chromosome Location:
Chr.19: 34677619 - 34686397 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07286930_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF302
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289181.1 NP_001276110.1 2 - 3 298 69
XM_017026978.1 XP_016882467.1 2 - 3 536 69
XM_017026979.1 XP_016882468.1 2 - 3 536 69
XM_017026980.1 XP_016882469.1 2 - 3 230 69
XM_017026981.1 XP_016882470.1 2 - 3 193 69
XM_017026983.1 XP_016882472.1 2 - 3 238 69
XM_017026984.1 XP_016882473.1 1 - 2 719 69
XM_017026985.1 XP_016882474.1 2 - 3 259 69
GenBank mRNA AK131278.1 - 2 - 3 220 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK131278.1, NM_001289181.1, XM_017026978.1, XM_017026979.1, XM_017026980.1, XM_017026981.1, XM_017026983.1, XM_017026984.1, XM_017026985.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0