This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07286953_m1
See other ENPP5 GE Assays ›
Gene Symbol:
ENPP5
Gene Name:
ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative)
Gene Aliases:
NPP-5, NPP5
Chromosome Location:
Chr.6: 46159182 - 46171045 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07286953_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ENPP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290072.1 NP_001277001.1 3 - 4 1124 78
NM_001290073.1 NP_001277002.1 2 - 3 771 78
NM_021572.5 NP_067547.1 2 - 3 1062 78
XM_005249260.3 XP_005249317.1 2 - 3 961 78
XM_011514786.2 XP_011513088.1 3 - 4 979 78
GenBank mRNA AK292842.1 - 2 - 3 1023 78
AK295192.1 - 2 - 3 732 78
AK312790.1 - 2 - 3 900 78
AY358514.1 - 3 - 4 1067 78
BC027615.2 - 2 - 3 962 4839342 78
BX647968.1 - 2 - 3 1033 78

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ENPP5

Gene Ontology Categories:

Function(s) Process(es)

cell communication
metabolic process
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline