This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07286954_m1
See other ENPP5 GE Assays ›
Gene Symbol:
ENPP5
Gene Name:
ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative)
Gene Aliases:
NPP-5, NPP5
Chromosome Location:
Chr.6: 46159182 - 46171045 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07286954_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ENPP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290073.1 NP_001277002.1 1 - 2 206 103
GenBank mRNA AK295192.1 - 1 - 2 167 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK295192.1, NM_001290073.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell communication
metabolic process
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0