This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07286968_m1
See other SDR39U1 GE Assays ›
Gene Symbol:
SDR39U1
Gene Name:
short chain dehydrogenase/reductase family 39U member 1
Gene Aliases:
C14orf124, HCDI
Chromosome Location:
Chr.14: 24439766 - 24442905 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07286968_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDR39U1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290293.2 NP_001277222.1 3 - 4 322 67
GenBank mRNA BX161384.1 - 2 - 3 273 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BX161384.1, NM_001290293.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

epimerase/racemase

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
oxidoreductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0