This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287021_m1
See other ZNF419 GE Assays ›
Gene Symbol:
ZNF419
Gene Name:
zinc finger protein 419
Gene Aliases:
ZAPHIR, ZNF419A
Chromosome Location:
Chr.19: 57487711 - 57494680 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287021_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF419
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098492.1 NP_001091962.1 1 - 2 277 93
NM_001098495.1 NP_001091965.1 1 - 2 277 93
NM_001291745.1 NP_001278674.1 1 - 2 277 93
GenBank mRNA DC315347.1 - 1 - 2 277 93
DC407547.1 - 1 - 2 235 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001098492.1, NM_001098495.1, NM_001291745.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0