This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287026_m1
See other IKZF1 GE Assays ›
Gene Symbol:
IKZF1
Gene Name:
IKAROS family zinc finger 1
Gene Aliases:
CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1
Chromosome Location:
Chr.7: 50303453 - 50405101 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287026_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IKZF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291845.1 NP_001278774.1 3 - 4 424 70
NM_001291846.1 NP_001278775.1 3 - 4 424 70
NM_001291847.1 NP_001278776.1 3 - 4 397 70
XM_011515058.2 XP_011513360.1 3 - 4 254 70
XM_011515059.2 XP_011513361.1 3 - 4 256 70
XM_011515060.2 XP_011513362.1 3 - 4 3404 70
XM_011515062.2 XP_011513364.1 3 - 4 534 70
XM_011515063.2 XP_011513365.1 3 - 4 451 70
XM_011515064.2 XP_011513366.1 3 - 4 4082 70
XM_011515065.2 XP_011513367.1 3 - 4 219 70
XM_011515066.2 XP_011513368.1 3 - 4 323 70
XM_011515067.2 XP_011513369.1 3 - 4 256 70
XM_011515071.2 XP_011513373.1 3 - 4 254 70
XM_011515073.2 XP_011513375.1 3 - 4 534 70
XM_017011667.1 XP_016867156.1 3 - 4 451 70
GenBank mRNA BM148203.1 - 3 - 4 416 70
BU537824.1 - 3 - 4 332 70
JX459579.1 - 3 - 4 324 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cell cycle
mesoderm development
covalent chromatin modification
lymphocyte differentiation
erythrocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-s2cfr:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0