This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287245_m1
See other NAT9 GE Assays ›
Gene Symbol:
NAT9
Gene Name:
N-acetyltransferase 9 (putative)
Gene Aliases:
EBSP, hNATL
Chromosome Location:
Chr.17: 74770529 - 74776425 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287245_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAT9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001305077.1 NP_001292006.1 4 - 5 501 84
NM_001305078.1 NP_001292007.1 4 - 5 500 84
NM_001305079.1 NP_001292008.1 4 - 5 504 84
NM_001305080.1 NP_001292009.1 4 - 5 504 84
NM_001305081.1 NP_001292010.1 4 - 5 501 84
NM_001305082.1 NP_001292011.1 4 - 5 649 84
NM_001305083.1 NP_001292012.1 4 - 5 501 84
NM_001305084.1 NP_001292013.1 4 - 5 504 84
NM_001305085.1 NP_001292014.1 4 - 5 501 84
NM_001305086.1 NP_001292015.1 4 - 5 501 84
NM_001305087.1 NP_001292016.1 4 - 5 524 84
NM_001305088.1 NP_001292017.1 3 - 4 415 84
NM_015654.4 NP_056469.2 4 - 5 504 84
XM_005257212.4 XP_005257269.1 3 - 4 343 84
XM_006721813.3 XP_006721876.1 3 - 4 791 84
XM_017024437.1 XP_016879926.1 4 - 5 430 84
XM_017024439.1 XP_016879928.1 4 - 5 471 84
XM_017024441.1 XP_016879930.1 3 - 4 803 84
GenBank mRNA AK225602.1 - 4 - 5 374 84
AK312958.1 - 4 - 5 374 84
AL050269.1 - 4 - 5 381 84
AL537925.3 - 4 - 5 581 84
AY632082.1 - 3 - 4 340 84
BC004195.2 - 4 - 5 410 3528894 84
BC004225.2 - 4 - 5 399 3538186 84
BG702300.1 - 4 - 5 437 84
BM909550.1 - 4 - 5 403 84
BP197633.1 - 4 - 5 416 84
BP231776.1 - 4 - 5 422 84
BQ068039.1 - 3 - 4 297 84
BU159218.1 - 4 - 5 399 84
BX333236.2 - 4 - 5 414 84
DA010490.1 - 4 - 5 444 84
DA259097.1 - 4 - 5 501 84
DB171198.1 - 4 - 5 418 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NAT9

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid acetylation
protein binding
N-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0