This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287247_m1
See other NAT9 GE Assays ›
Gene Symbol:
NAT9
Gene Name:
N-acetyltransferase 9 (putative)
Gene Aliases:
EBSP, hNATL
Chromosome Location:
Chr.17: 74770529 - 74776425 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287247_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAT9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001305079.1 NP_001292008.1 6 - 7 584 92
NM_001305088.1 NP_001292017.1 5 - 6 495 92
XM_017024439.1 XP_016879928.1 6 - 7 551 92
GenBank mRNA BP197633.1 - 6 - 7 496 92
BQ068039.1 - 5 - 6 377 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid acetylation
protein binding
N-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6s775:80/100.66.79.102:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0