This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287261_m1
See other KAT6A GE Assays ›
Gene Symbol:
KAT6A
Gene Name:
lysine acetyltransferase 6A
Gene Aliases:
MOZ, MRD32, MYST-3, MYST3, RUNXBP2, ZC2HC6A, ZNF220
Chromosome Location:
Chr.8: 41929479 - 42052026 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287261_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KAT6A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001305878.1 NP_001292807.1 11 - 12 2358 94
NM_006766.4 NP_006757.2 11 - 12 2358 94
XM_011544656.2 XP_011542958.1 13 - 14 2584 94
XM_011544657.2 XP_011542959.1 12 - 13 2451 94
XM_011544658.2 XP_011542960.1 13 - 14 2540 94
XM_011544659.2 XP_011542961.1 13 - 14 2563 94
XM_017013863.1 XP_016869352.1 12 - 13 2452 94
XM_017013864.1 XP_016869353.1 12 - 13 2408 94
GenBank mRNA AK128323.1 - 4 - 5 1740 94
BC142659.1 - 11 - 12 2305 94
BC142959.1 - 11 - 12 2319 94
U47742.1 - 11 - 12 2300 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KAT6A

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

DNA packaging
nucleosome assembly
transcription, DNA-templated
protein acetylation
histone acetylation
myeloid cell differentiation
histone H3 acetylation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
cellular senescence
regulation of signal transduction by p53 class mediator
DNA binding
transcription coactivator activity
histone acetyltransferase activity
protein binding
transcription factor binding
zinc ion binding
acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0