This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287279_m1
See other EIF3M GE Assays ›
Gene Symbol:
EIF3M
Gene Name:
eukaryotic translation initiation factor 3 subunit M
Gene Aliases:
B5, GA17, PCID1, TANGO7, hfl-B5
Chromosome Location:
Chr.11: 32583767 - 32602873 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287279_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3M
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001307929.1 NP_001294858.1 1 - 2 166 68
GenBank mRNA AK001768.1 - 1 - 2 102 68
AK304378.1 - 1 - 2 102 68
AV713889.1 - 1 - 2 83 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
cytoplasmic translational initiation
translational initiation
regulation of translational initiation
translation initiation factor activity
protein binding
translation initiation factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0