This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287281_m1
See other EIF3M GE Assays ›
Gene Symbol:
EIF3M
Gene Name:
eukaryotic translation initiation factor 3 subunit M
Gene Aliases:
B5, GA17, PCID1, TANGO7, hfl-B5
Chromosome Location:
Chr.11: 32583767 - 32602873 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287281_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3M
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001307929.1 NP_001294858.1 5 - 6 521 102
NM_006360.5 NP_006351.2 8 - 9 917 102
XM_011519840.2 XP_011518142.1 8 - 9 941 102
GenBank mRNA AF064603.1 - 8 - 9 847 102
AF277183.1 - 4 - 5 286 102
AK001768.1 - 4 - 5 1422 102
AK131064.1 - 8 - 9 871 102
AK222564.1 - 8 - 9 836 102
AK292139.1 - 8 - 9 859 102
AK304378.1 - 5 - 6 457 102
AK307121.1 - 5 - 6 1537 102
AK312512.1 - 8 - 9 886 102
AV713889.1 - 5 - 6 438 102
AY769947.1 - 8 - 9 796 102
BC019103.2 - 8 - 9 839 5021401 102
BC020458.1 - 4 - 5 308 102
BC051292.1 - 8 - 9 841 3607828 102
CR450300.1 - 8 - 9 796 102

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF3M

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
cytoplasmic translational initiation
translational initiation
regulation of translational initiation
translation initiation factor activity
protein binding
translation initiation factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0