This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287313_g1
See other CIAPIN1 GE Assays ›
Gene Symbol:
CIAPIN1
Gene Name:
cytokine induced apoptosis inhibitor 1
Gene Aliases:
Anamorsin, DRE2, PRO0915
Chromosome Location:
Chr.16: 57428169 - 57447528 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287313_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CIAPIN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308347.1 NP_001295276.1 4 - 5 584 103
NM_001308358.1 NP_001295287.1 4 - 5 623 103
NM_020313.3 NP_064709.2 4 - 5 623 103
XM_005256063.4 XP_005256120.1 4 - 5 475 103
GenBank mRNA AF116609.1 - 2 - 3 234 103
AF248964.1 - 4 - 5 527 103
AK074263.1 - 4 - 5 464 103
AK292281.1 - 4 - 5 427 103
AK295024.1 - 3 - 4 856 103
AL136613.1 - 4 - 5 445 103
BC002568.2 - 4 - 5 465 3139226 103
BC024196.2 - 4 - 5 504 4816594 103
BC067303.1 - 4 - 5 623 30389410 103
BC071740.1 - 4 - 5 498 5563094 103
BG746807.1 - 1 469 103
BJ991015.1 - 4 - 5 504 103
CR533545.1 - 3 - 4 342 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF116609.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CIAPIN1

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
iron-sulfur cluster assembly
hemopoiesis
methylation
negative regulation of apoptotic process
protein binding
methyltransferase activity
electron carrier activity
metal ion binding
2 iron, 2 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0