This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287315_g1
See other CIAPIN1 GE Assays ›
Gene Symbol:
CIAPIN1
Gene Name:
cytokine induced apoptosis inhibitor 1
Gene Aliases:
Anamorsin, DRE2, PRO0915
Chromosome Location:
Chr.16: 57428169 - 57447528 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287315_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CIAPIN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308347.1 NP_001295276.1 7 - 8 952 105
NM_020313.3 NP_064709.2 7 - 8 991 105
GenBank mRNA AF116609.1 - 5 - 6 602 105
AF248964.1 - 7 - 8 895 105
AK074263.1 - 7 - 8 832 105
AK292281.1 - 7 - 8 795 105
AL136613.1 - 7 - 8 813 105
BC002568.2 - 7 - 8 833 3139226 105
BC024196.2 - 7 - 8 872 4816594 105
BC067303.1 - 7 - 8 991 30389410 105
BC071740.1 - 7 - 8 866 5563094 105
CR533545.1 - 6 - 7 710 105

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CIAPIN1

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
iron-sulfur cluster assembly
hemopoiesis
methylation
negative regulation of apoptotic process
protein binding
methyltransferase activity
electron carrier activity
metal ion binding
2 iron, 2 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline