This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287317_m1
See other RAMP1 GE Assays ›
Gene Symbol:
RAMP1
Gene Name:
receptor activity modifying protein 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 237858714 - 237912117 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287317_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAMP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308353.1 NP_001295282.1 2 - 3 246 55
XM_017003154.1 XP_016858643.1 1 - 2 192 55
GenBank mRNA BI548986.1 - 2 - 3 172 55

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001308353.1, XM_017003154.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
calcium ion transport
intracellular protein transport
G-protein coupled receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
protein transport
positive regulation of cAMP biosynthetic process
receptor internalization
positive regulation of protein glycosylation
protein localization to plasma membrane
receptor activity
calcitonin receptor activity
protein binding
protein transporter activity
coreceptor activity
calcitonin receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bs78j:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0