This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287321_m1
See other NME6 GE Assays ›
Gene Symbol:
NME6
Gene Name:
NME/NM23 nucleoside diphosphate kinase 6
Gene Aliases:
IPIA-ALPHA, NDK 6, NM23-H6
Chromosome Location:
Chr.3: 48289369 - 48302259 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287321_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NME6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308426.1 NP_001295355.1 1 - 2 332 89
NM_001308431.1 NP_001295360.1 1 - 2 332 89
NM_001308435.1 NP_001295364.1 1 - 2 332 89
GenBank mRNA CT002144.1 - 1 - 2 332 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001308426.1, NM_001308431.1, NM_001308435.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

kinase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleoside diphosphate phosphorylation
GTP biosynthetic process
UTP biosynthetic process
CTP biosynthetic process
apoptotic process
negative regulation of cell growth
negative regulation of mitotic nuclear division
nucleoside diphosphate kinase activity
ATP binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline