This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287376_g1
See other RAC3 GE Assays ›
Gene Symbol:
RAC3
Gene Name:
ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3)
Gene Aliases:
-
Chromosome Location:
Chr.17: 82031656 - 82034204 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287376_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001316307.1 NP_001303236.1 5 - 6 557 100
GenBank mRNA BI668056.1 - 5 - 6 511 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
regulation of neuron maturation
cerebral cortex GABAergic interneuron development
cell projection assembly
actin cytoskeleton organization
neuron projection development
positive regulation of cell adhesion mediated by integrin
intracellular signal transduction
homeostasis of number of cells within a tissue
neuromuscular process controlling balance
regulation of small GTPase mediated signal transduction
synaptic transmission, GABAergic
positive regulation of substrate adhesion-dependent cell spreading
GTPase activity
protein binding
GTP binding
calcium-dependent protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0