This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287377_g1
See other POLR2E GE Assays ›
Gene Symbol:
POLR2E
Gene Name:
RNA polymerase II subunit E
Gene Aliases:
RPABC1, RPB5, XAP4, hRPB25, hsRPB5
Chromosome Location:
Chr.19: 1086579 - 1095392 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287377_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR2E
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001316323.1 NP_001303252.1 3 - 4 330 101
NM_001316324.1 NP_001303253.1 4 - 5 478 101
NM_002695.3 NP_002686.2 4 - 5 505 101
XM_011528070.2 XP_011526372.1 4 - 5 500 101
XM_017026883.1 XP_016882372.1 3 - 4 319 101
GenBank mRNA AK122813.1 - 3 - 4 1667 101
AK295975.1 - 4 - 5 423 101
AK312625.1 - 4 - 5 458 101
BC004441.1 - 4 - 5 450 2820565 101
BC034144.1 - 4 - 5 470 4617000 101
DC351659.1 - 3 - 4 313 101
DC414435.1 - 4 - 5 506 101
J04965.1 - 4 - 5 452 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLR2E

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
transcription-coupled nucleotide-excision repair
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
transcription from RNA polymerase III promoter
fibroblast growth factor receptor signaling pathway
gene expression
viral process
gene silencing by RNA
positive regulation of type I interferon production
somatic stem cell population maintenance
snRNA transcription from RNA polymerase II promoter
positive regulation of gene expression, epigenetic
positive regulation of viral transcription
RNA polymerase I activity
RNA polymerase II activity
RNA polymerase III activity
DNA binding
DNA-directed RNA polymerase activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0