This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287464_m1
See other FAHD1 GE Assays ›
Gene Symbol:
FAHD1
Gene Name:
fumarylacetoacetate hydrolase domain containing 1
Gene Aliases:
C16orf36, YISKL
Chromosome Location:
Chr.16: 1827224 - 1840207 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287464_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAHD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001018104.2 NP_001018114.1 2 - 3 759 66

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FAHD1

Panther Classification:

Molecular Function -

hydrolase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
oxaloacetate decarboxylase activity
acetylpyruvate hydrolase activity
fumarylpyruvate hydrolase activity
metal ion binding
acylpyruvate hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5cqgw:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0