This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287621_g1
See other BET1 GE Assays ›
Gene Symbol:
BET1
Gene Name:
Bet1 golgi vesicular membrane trafficking protein
Gene Aliases:
HBET1
Chromosome Location:
Chr.7: 93962770 - 94004382 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287621_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BET1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317739.1 NP_001304668.1 1 - 2 187 101
NM_005868.5 NP_005859.1 1 - 2 187 101
NR_133908.1 1 - 2 187 101
NR_133909.1 1 - 2 187 101
GenBank mRNA AF007551.1 - 1 - 2 140 101
AK313984.1 - 1 - 2 160 101
BC000899.1 - 1 - 2 76 2899921 101
BC012595.1 - 1 - 2 170 4096064 101
BG721261.1 - 1 - 2 187 101
BX505568.1 - 1 - 2 187 101
CR749267.1 - 1 - 2 138 101
DA127369.1 - 1 - 2 160 101

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF007551.1, AK313984.1, BC000899.1, BC012595.1, CR749267.1, NM_001317739.1, NM_005868.5 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BET1

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

ER to Golgi vesicle-mediated transport
protein transport
COPII vesicle coating
vesicle fusion with Golgi apparatus
protein binding
syntaxin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0