Assay Details
| Gene Symbol: SLC10A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317818.1 | NP_001304747.1 | 2 - 3 | 430 | 93 | |
| GenBank mRNA | BC023288.1 | - | 2 - 3 | 381 | 93 | |
More Information
Set Membership: |
|

SLC10A7
Genomic Map