This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287709_m1
See other TRAPPC4 GE Assays ›
Gene Symbol:
TRAPPC4
Gene Name:
trafficking protein particle complex 4
Gene Aliases:
CGI-104, HSPC172, PTD009, SBDN, SYNBINDIN, TRS23
Chromosome Location:
Chr.11: 119018469 - 119023674 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287709_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAPPC4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318486.1 NP_001305415.1 3 - 4 796 80
NM_001318489.1 NP_001305418.1 4 - 5 738 80
NM_001318490.1 NP_001305419.1 4 - 5 771 80
NM_001318492.1 NP_001305421.1 4 - 5 1007 80
NM_001318494.1 NP_001305423.1 2 - 3 621 80
NM_016146.5 NP_057230.1 4 - 5 900 80
GenBank mRNA AF078862.1 - 4 - 5 830 80
AF105025.1 - 4 - 5 838 80
AF151862.1 - 4 - 5 836 80
AF161520.1 - 4 - 5 623 80
AK290520.1 - 4 - 5 603 80
AK297422.1 - 4 - 5 441 80
BC010866.1 - 4 - 5 599 3876778 80
BF971819.1 - 4 - 5 686 80
BP246594.1 - 3 - 4 485 80
BU624012.1 - 1 - 2 379 80

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRAPPC4

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

ER to Golgi vesicle-mediated transport
dendrite development
positive regulation of GTPase activity
COPII vesicle coating
protein binding
Rab guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0