This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287713_g1
See other MSANTD4 GE Assays ›
Gene Symbol:
MSANTD4
Gene Name:
Myb/SANT DNA binding domain containing 4 with coiled-coils
Gene Aliases:
KIAA1826
Chromosome Location:
Chr.11: 106007920 - 106022287 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287713_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSANTD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318747.1 NP_001305676.1 2 - 3 1060 84
NM_001318748.1 NP_001305677.1 2 - 3 1056 84
NM_001318749.1 NP_001305678.1 2 - 3 774 84
NM_001318750.1 NP_001305679.1 2 - 3 701 84
NM_032424.2 NP_115800.1 2 - 3 1934 84
XM_011543022.2 XP_011541324.1 3 - 4 1962 84
XM_017018416.1 XP_016873905.1 3 - 4 1072 84
XM_017018417.1 XP_016873906.1 2 - 3 659 84
GenBank mRNA AB058729.1 - 2 - 3 1874 84
AF253976.1 - 1 - 2 124 84
AK027790.1 - 1 - 2 94 84
AK293050.1 - 2 - 3 730 84
AL834523.1 - 2 - 3 624 84
BC041118.1 - 2 - 3 631 5526792 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK027790.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MSANTD4

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0