This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287751_g1
See other RPL34 GE Assays ›
Gene Symbol:
RPL34
Gene Name:
ribosomal protein L34
Gene Aliases:
L34
Chromosome Location:
Chr.4: 108620558 - 108630484 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287751_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL34
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001319235.1 NP_001306164.1 1 - 2 123 95
NM_033625.3 NP_296374.1 1 - 2 123 95
GenBank mRNA BC001773.1 - 1 - 2 57 3355603 95
BE394007.1 - 1 - 2 89 95
BU588998.1 - 1 - 2 123 95
CD175155.1 - 1 - 2 145 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC001773.1, NM_001319235.1, NM_033625.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPL34

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
ribosome biogenesis
cell-cell adhesion
RNA binding
structural constituent of ribosome
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline