This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287831_g1
See other PCNP GE Assays ›
Gene Symbol:
PCNP
Gene Name:
PEST proteolytic signal containing nuclear protein
Gene Aliases:
-
Chromosome Location:
Chr.3: 101574094 - 101594465 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287831_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCNP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320401.1 NP_001307330.1 2 - 3 406 70
NR_135230.1 2 - 3 354 70
GenBank mRNA DA710370.1 - 2 - 3 296 70
DB481581.1 - 2 - 3 242 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline