This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287853_g1
See other SAT2 GE Assays ›
Gene Symbol:
SAT2
Gene Name:
spermidine/spermine N1-acetyltransferase family member 2
Gene Aliases:
SSAT2
Chromosome Location:
Chr.17: 7626234 - 7627878 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287853_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320845.1 NP_001307774.1 2 - 3 375 86
NM_001320846.1 NP_001307775.1 2 - 3 375 86
XM_017024073.1 XP_016879562.1 2 - 3 375 86
XM_017024074.1 XP_016879563.1 2 - 3 377 86
GenBank mRNA BC095466.1 - 2 - 3 313 86
BP317593.1 - 2 - 3 376 86
HY155008.1 - 2 - 3 361 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

putrescine catabolic process
spermidine acetylation
spermine acetylation
putrescine acetylation
nor-spermidine metabolic process
diamine N-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0