This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287903_g1
See other COX11 GE Assays ›
Gene Symbol:
COX11
Gene Name:
COX11, cytochrome c oxidase copper chaperone
Gene Aliases:
COX11P
Chromosome Location:
Chr.17: 54951898 - 54971024 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287903_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COX11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162861.2 NP_001156333.1 2 - 3 660 63
NM_001162862.2 NP_001156334.1 2 - 3 660 63
NM_001321518.1 NP_001308447.1 2 - 3 660 63
NM_004375.4 NP_004366.1 2 - 3 660 63
NR_027941.2 2 - 3 660 63
NR_027942.2 2 - 3 660 63
NR_135677.1 2 - 3 660 63
XM_011524342.2 XP_011522644.1 2 - 3 2899 63
XM_017024192.1 XP_016879681.1 2 - 3 2899 63
XM_017024193.1 XP_016879682.1 2 - 3 2899 63
XM_017024194.1 XP_016879683.1 2 - 3 655 63
XM_017024195.1 XP_016879684.1 2 - 3 655 63
XM_017024196.1 XP_016879685.1 2 - 3 655 63
GenBank mRNA AF044321.1 - 2 - 3 536 63
AI828544.1 - 3 - 4 529 63
AK293851.1 - 2 - 3 493 63
AK295381.1 - 2 - 3 578 63
BC005895.1 - 2 - 3 531 4282744 63
BM975167.1 - 3 - 4 562 63
CR541837.1 - 2 - 3 521 63
U79270.1 - 1 - 2 151 63

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene COX11

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

respiratory gaseous exchange
respiratory chain complex IV assembly
aerobic respiration
negative regulation of glucokinase activity
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
copper ion binding
electron carrier activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0