This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287947_m1
See other ZSCAN5A GE Assays ›
Gene Symbol:
ZSCAN5A
Gene Name:
zinc finger and SCAN domain containing 5A
Gene Aliases:
ZNF495, ZSCAN5
Chromosome Location:
Chr.19: 56221303 - 56368383 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287947_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZSCAN5A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322061.1 NP_001308990.1 3 - 4 533 64
NM_001322062.1 NP_001308991.1 2 - 3 301 64
XM_005259254.1 XP_005259311.1 2 - 3 301 64
XM_006723377.3 XP_006723440.1 1 - 2 585 64
XM_017027298.1 XP_016882787.1 1 - 2 570 64
GenBank mRNA AK098700.1 - 1 - 2 58 64
DB028988.1 - 2 - 3 301 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK098700.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0