This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287980_g1
See other DSE GE Assays ›
Gene Symbol:
DSE
Gene Name:
dermatan sulfate epimerase
Gene Aliases:
DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2
Chromosome Location:
Chr.6: 116254152 - 116441261 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287980_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DSE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080976.2 NP_001074445.1 5 - 6 1367 108
NM_001322937.1 NP_001309866.1 6 - 7 1923 108
NM_001322938.1 NP_001309867.1 6 - 7 1835 108
NM_001322939.1 NP_001309868.1 5 - 6 1250 108
NM_001322940.1 NP_001309869.1 6 - 7 1919 108
NM_001322941.1 NP_001309870.1 5 - 6 1417 108
NM_013352.3 NP_037484.1 5 - 6 1421 108
XM_017010795.1 XP_016866284.1 7 - 8 1556 108
XM_017010796.1 XP_016866285.1 4 - 5 1177 108
GenBank mRNA AF098066.1 - 5 - 6 1265 108
AK128607.1 - 1 - 2 1019 108
AK301513.1 - 5 - 6 1250 108
BC039245.1 - 5 - 6 1312 5272885 108

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DSE

Gene Ontology Categories:

Function(s) Process(es)

heparan sulfate proteoglycan biosynthetic process
chondroitin sulfate biosynthetic process
dermatan sulfate biosynthetic process
chondroitin-glucuronate 5-epimerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline