This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288061_m1
See other ZNF239 GE Assays ›
Gene Symbol:
ZNF239
Gene Name:
zinc finger protein 239
Gene Aliases:
HOK-2, MOK2
Chromosome Location:
Chr.10: 43554516 - 43574619 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288061_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF239
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001324352.1 NP_001311281.1 3 - 4 245 96
XM_011540234.2 XP_011538536.1 2 - 3 205 96
XM_011540235.2 XP_011538537.1 2 - 3 204 96
XM_011540236.2 XP_011538538.1 2 - 3 213 96
XM_011540237.2 XP_011538539.1 2 - 3 163 96
XM_017016740.1 XP_016872229.1 3 - 4 242 96
GenBank mRNA DA597035.1 - 3 - 4 224 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
RNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0