This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288143_m1
See other NAA38 GE Assays ›
Gene Symbol:
NAA38
Gene Name:
N(alpha)-acetyltransferase 38, NatC auxiliary subunit
Gene Aliases:
LSMD1, PFAAP2
Chromosome Location:
Chr.17: 7856685 - 7885465 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288143_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAA38
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011524037.2 XP_011522339.1 3 - 4 287 105
XM_017025225.1 XP_016880714.1 3 - 4 194 105
XM_017025226.1 XP_016880715.1 3 - 4 280 105

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011524037.2, XM_017025225.1, XM_017025226.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of apoptotic process
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0